Invitrogen™ RNase-free Tips DFS Item
Aerosol filter prevents cross-contamination of samples
$162.00 - $240.00
Specifications
Includes: Ten racks containing 8 x 12 tips each
Products 4
Catalog Number Mfr. No. Volume (Metric) Compatibility Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Compatibility Price Quantity  
AM12645 View Documents Invitrogen™
LSAM12645
20μL Compatible with Oxford Benchmnate/Nichiryo pipetters Each for $240.00
AM12648 View Documents Invitrogen™
LSAM12648
100μL Compatible with Oxford Benchmnate/Nichiryo pipetters Each for $236.00
AM12655 View Documents Invitrogen™
LSAM12655
200μL Not compatible with Oxford Benchmnate/Nichiryo pipetters Each for $236.00
AM12650 View Documents Invitrogen™
LSAM12650
200μL Compatible with Oxford Benchmnate/Nichiryo pipetters Each for $162.00
Description
Description
Compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipetters.
  • Tips are RNase- and DNase-free and ideal for handling RNA, PCR applications and working with radioisotopes or toxic chemicals
  • Each tip lot is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, PCR, PCR and Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Total RNA from Animal Cells and Tissues, Transcriptome RNA Isolation, mRNA Isolation, microRNA Isolation
Specifications
Specifications
0.0
icon-pdf.png
For Research Use Only. Not for use in diagnostic procedures.