Invitrogen™ Barrier (Filter) Tips, 200 μL size
Barrier tips prevent cross-contamination of samples by means of an aerosol filter.
Supplier: Invitrogen™ AM12655
Description
Compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipetters.
- Tips are RNase- and DNase-free and ideal for handling RNA, PCR applications and working with radioisotopes or toxic chemicals
- Each tip lot is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
Specifications
200 μL | |
Sterile | |
Rack | |
Store at room temperature. | |
Universal Fit |
Filtered | |
Autoclavable | |
Universal Fit | |
Room Temperature | |
Invitrogen |
For Research Use Only. Not for use in diagnostic procedures.