Please login to your online account to display your discounted pricing

Ambion™ ArrayScript™ Reverse Transcriptase DFS Item


Suitable for RT and RT-PCR applications where high yields of full-length cDNA are required, such as cDNA library construction, 5' RLM-RACE, etc.

Manufacturer: life technologies  LSAM2048

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM2048

  • / Each
 

Description & Specifications

Specifications

Temperature (English) Operating 98.6deg.F
Sample Type ssRNA, ssDNA, RNA-DNA Hybrid
Storage Requirements Store at -20deg.C
Includes 4000 units supplied with a 10X Reaction Buffer
Quantity 4000 units
For Use With (Application) RT and RT-PCR
Temperature (Metric) Operating 37deg.C