missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ ArrayScript™ Reverse Transcriptase DFS Item

Suitable for RT and RT-PCR applications where high yields of full-length cDNA are required, such as cDNA library construction, 5' RLM-RACE, etc.

$163.00 - $163.00

Specifications

For Use With (Application) RT and RT-PCR
Sample Type ssRNA, ssDNA, RNA-DNA Hybrid
Storage Requirements Store at -20deg.C
Temperature (English) Operating 98.6deg.F
Temperature (Metric) Operating 37deg.C
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Price Quantity  
Catalog Number Mfr. No. Includes Quantity Price Quantity  
AM2048 Life Technologies
LSAM2048
4000 units supplied with a 10X Reaction Buffer 4000 units Each for $163.00
Specifications

Description

  • Ambion ArrayScript Reverse Transcriptase is a M-MLV reverse transcriptase that has been engineered to produce high yields of full-length cDNA
  • 4000U are supplied along with a 10X Reaction Buffer. Reverse transcriptases (RTs) are used in a variety of molecular biology experiments where success can depend upon full-length cDNA synthesized during the reverse transcription reaction
  • In RNA amplification experiments, up to twice as much cRNA is synthesized when the first-strand cDNA is generated with ArrayScript RT compared to wild type M-MLV, AMV, or other engineered M-MLV reverse transcriptases when low amounts of total RNA (∽100ng) are used. This can eliminate the need to perform two rounds of cRNA amplification when working with limiting amounts of total RNA

Optimal reaction temperature: 37°C
Final product size: 7kb or less

Applications:

PCR & Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

RT and RT-PCR
Store at -20deg.C
37deg.C
ssRNA, ssDNA, RNA-DNA Hybrid
98.6deg.F