Invitrogen™ Ambion™ ANTI-RNase (15-30U/μL) DFS Item
More robust RNase interaction over human placental ribonuclease inhibitor
Manufacturer:  Invitrogen™ LSAM2690
 View more versions of this product
Catalog No. AM2690
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
15 to 30U/μL
ANTI-RNase
Store at -20°C. Do not store in a frost–free freezer or at -70°C.
Non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes
2500 units