missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ ANTI-RNase (15-30U/μL) DFS Item

More robust RNase interaction over human placental ribonuclease inhibitor

$133.35 - $450.00

Specifications

Concentration 15 to 30U/μL
Product Type ANTI-RNase
For Use With (Application) Non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes
Storage Requirements Store at -20°C. Do not store in a frost–free freezer or at -70°C.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM2690 Invitrogen™
LSAM2690
2500 units Each for $133.35
AM2692 Invitrogen™
LSAM2692
10,000 units Each for $450.00
Specifications

Description

Ambion ANTI-RNase is a protein ribonuclease inhibitor that non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes. No inhibition is seen with RNase 1, T1, T2, H, U1, U2, or CL3. ANTI-RNase is distinct from human placental ribonuclease inhibitor in that it has a more robust interaction with RNase and does not release active RNase in the absence of DTT or other reducing agents. ANTI-RNase may be used in any application where placental ribonuclease inhibitor is used. It is ideal for use in invitro transcription and translation, cDNA synthesis PCR, RT-PCR, RNA isolation procedures, and the preparation of RNase-free antibodies. ANTI-RNase is added to reactions to give a final concentration of 1U/μL. Slight inhibition of ANTI-RNase is measured in the presence of DTT (≤20mM). ANTI-RNase is thoroughly tested for contaminating RNase, exonuclease, and protease activity.

Unit Definition: One unit is the amount of protein required to inhibit the activity of 5ng RNase A by 50%.

Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Condition: Dry ice

Description & Specifications

Specifications

15 to 30U/μL
ANTI-RNase
Non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes
Store at -20°C. Do not store in a frost–free freezer or at -70°C.