Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ ANTI-RNase (15-30U/μL) DFS Item


More robust RNase interaction over human placental ribonuclease inhibitor

Manufacturer: invitrogen™  LSAM2692

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM2692

  • / Each
 

Description & Specifications

Specifications

Concentration 15 to 30U/μL
For Use With (Application) Non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes
Volume (Metric) 20 U/μL
Quantity 10,000 units
Product Type ANTI-RNase
Quantity 10,000 units
Storage Requirements Store at -20°C. Do not store in a frost–free freezer or at -70°C.