Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Agarose LE™, General Use DFS Item


Prepare gels with concentrations as low as 0.5%

Manufacturer: invitrogen™  LSAM9042

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM9042

  • / Each
 

Description & Specifications

Specifications

Quantity 500g
Storage Requirements Store at room temperature.
For Use With (Application) Gel Electrophoresis