missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Agarose LE™, General Use DFS Item

Prepare gels with concentrations as low as 0.5%

$842.00 - $842.00

Specifications

For Use With (Application) Gel Electrophoresis
Storage Requirements Store at room temperature.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM9042 View Documents Invitrogen™
LSAM9042
500g Each for $842.00
Description

Description

Ambion Agarose-LE™ (General Use) is a high-purity, functionally tested, general use agarose suitable for routine nucleic acid electrophoresis. It has a low electroendosmosis value (EEO -mr 0.103) ensuring high electrophoretic mobility of nucleic acids. Because of the high gel strength, easy-to-handle gels can be prepared with this agarose, at concentrations as low as 0.5%. This can improve the separation of larger DNA fragments (10 to 20 kb) over conventional electrophoresis techniques.

  • Separation Range: 100bp to >30kb

Accessory Product:

  • DNA can be rapidly recovered from this agarose with Spin Columns (Cat. No. AM10065) and is compatible with restriction endonuclease digestion and ligation.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Gel Electrophoresis
Store at room temperature.

For Research Use Only. Not for use in diagnostic procedures.