Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Acrylamide/Bis 19:1, 40% (w/v) solution DFS Item


Ready-to-use solution reduces acrylamide and bis-acrylamide powder contact hazards

Manufacturer: invitrogen™  LSAM9022

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM9022

  • / Each
 

Description & Specifications

Specifications

Quantity 500mL
Storage Requirements Store at 4°C.