missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ Acrylamide/Bis 19:1, 40% (w/v) solution DFS Item

Ready-to-use solution reduces acrylamide and bis-acrylamide powder contact hazards

$147.00 - $232.00

Specifications

Storage Requirements Store at 4°C.
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AM9022 Invitrogen™
LSAM9022
500mL Each for $147.00
AM9024 Invitrogen™
LSAM9024
2 x 500mL Each for $232.00
Specifications

Description

Ambion Acrylamide/Bis 19:1 is a 40% (w/v) solution of acrylamide (38%) and bis-acrylamide (2%) ideal for use in ribonuclease protection assay, sequencing gels, and sizing DNA or RNA fragments. The solution is provided in a ready-to-use form, reducing the dust, inhalation, and contact hazard associated with weighing and preparing acrylamide and bis-acrylamide powders and solutions. Concentrations are carefully controlled and all solutions are micro-filtered prior to packaging. All Ambion acrylamides are rigorously tested for contaminating non-specific endonuclease, exonuclease, and RNase activity, and for functionality.

Acrylamide Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

Description & Specifications

Specifications

Store at 4°C.