Please login to your online account to display your discounted pricing

Amberlyst™ A26, OH-form, ion-exchange resin, ACROS Organics™


Quantity 100mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC202181000

 View more versions of this product

Catalog No. AC202181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 100mL
Fieser 01,511; 02,227; 04,266; 05,355; 06,302; 07,182; 08,263
Merck Index 10, 381
Chemical Name or Material Amberlyst™ A26
CAS 39339-85-0
MDL Number MFCD00145579
Packaging Plastic bottle

Specifications

Chemical Name or Material Amberlyst™ A26
CAS 39339-85-0
MDL Number MFCD00145579