Alfa Aesar™ Amberlyst™ 15(H), wet, ion exchange resin
Manufacturer:  Alfa Aesar™ 44079A7
 View more versions of this product
Catalog No. AA44079A7
Specifications
Specifications
Amberlyst™ 15 (H) ion exchange resin
39389-20-3
MFCD00145841
Insoluble in water.
SIWVGXQOXWGJCI-UHFFFAOYSA-N
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
314.399
Beads
Wet, Styrene-DVB; Strongly acidic macroreticular resin
C18H18O3S
1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
5kg
Hygroscopic
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
170197
Chemical Identifiers
39389-20-3
314.399
SIWVGXQOXWGJCI-UHFFFAOYSA-N
170197
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
C18H18O3S
MFCD00145841
1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
Safety and Handling
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
Documents