Amberlyst™ 15(H), Pale brown beads, Alfa Aesar™
Strongly acidic macroreticular resin
Manufacturer:  Alfa Aesar 8907918
 View more versions of this product
Catalog No. AA8907918
Specifications
Specifications
Amberlyst™ 15(H)
Odorless
MFCD00145841
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
SIWVGXQOXWGJCI-UHFFFAOYSA-N
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
170197
39389-20-3
C18H18O3S
Brown
50g
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
314.399
Beads
Chemical Identifiers
39389-20-3
314.399
SIWVGXQOXWGJCI-UHFFFAOYSA-N
170197
C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O
C18H18O3S
MFCD00145841
amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
SDS Documents