Learn More
Amberlite XAD-7, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1956436
Description
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.
Solubility
Insoluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Amberlite XAD-7 | |
MFCD00132705 | |
500 g |
37380-43-1 | |
14,382 | |
Insoluble in water. |
Chemical Identifiers
37380-43-1 | |
MFCD00132705 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only