Please login to your online account to display your discounted pricing

Amberlite™ XAD-4, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC202230025

 View more versions of this product

Catalog No. AC202230025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Quantity 2.5kg
Color White to almost white
Particle Size 2.0 (uniformity coefficient) Max. 0.490 to 0.690mm (harmonic mean size) 5.0% Max. (<0.300mm) 5.0% Max. (>1.180 mm)
Packaging Plastic bottle
MDL Number MFCD00132704
Moisture Holding Capacity moisture holding capacity: 54.0 to 60.0%
Sodium Chloride 20.0 to 32.0 g/l
Chemical Name or Material Amberlite™ XAD-4
Additional Information Ponceau dye test: < 1.0
CAS 9003-69-4

Specifications

Chemical Name or Material Amberlite™ XAD-4
CAS 9003-69-4
MDL Number MFCD00132704