missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite XAD-16

$85.74 - $232.43

Chemical Identifiers

CAS 104219-63-8
MDL Number MFCD00145831
Merck Index 14,382
Chemical Name or Material Amberlite XAD-16
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1956536 SDS Alfa Aesar™
L1956536*
500g Each for $232.43
AAL1956522 SDS Alfa Aesar™
L1956522*
100g Each for $85.74
Specifications

Description

Applications
Amberlite XAD-16 is a polymeric adsorbent widely utilized for adsorption of organic substances from aqueous systems and polar solvents. It is useful for the removal of detergents from protein solutions. It is also used in the recovery and purification of antibiotics, water soluble steroids, amino acids and proteins. It plays a vital role in the removal of non polar compounds from polar solvents.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.
Description & Specifications

Chemical Identifiers

104219-63-8
MFCD00145831
14,382
Amberlite XAD-16

Specifications

104219-63-8
14,382
Amberlite XAD-16
MFCD00145831
500g