Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite XAD-16


Quantity 500g

Manufacturer: alfa aesar™  L1956536*

 View more versions of this product

Catalog No. AAL1956536

  • / Each
 

Description & Specifications

Chemical Identifiers

CAS 104219-63-8
Chemical Name or Material Amberlite XAD-16
MDL Number MFCD00145831
Merck Index 14,382
Quantity 500g

Specifications

Chemical Name or Material Amberlite XAD-16
CAS 104219-63-8
MDL Number MFCD00145831
Merck Index 14,382

Applications
Amberlite XAD-16 is a polymeric adsorbent widely utilized for adsorption of organic substances from aqueous systems and polar solvents. It is useful for the removal of detergents from protein solutions. It is also used in the recovery and purification of antibiotics, water soluble steroids, amino acids and proteins. It plays a vital role in the removal of non polar compounds from polar solvents.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.