missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amberlite™ IRP-69 ion-exchange resin, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 301315000

 View more versions of this product

Catalog No. AC301315000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

500g
9.4 to 11.5% (Na)
Particle size analysis: screens on 100 Mesh: <1%, screens on 200 Mesh: 10.0 to 25.0%
10% max. (K.F.)
MFCD00132786
potassium exchange capacity: 110 to 135mg/g
14, 382
Powder
ammonium salts: Passes Test
55464-99-8
Plastic bottle
Amberlite™ IRP-69

Chemical Identifiers

MFCD00132786
Amberlite™ IRP-69
9.4 to 11.5% (Na)