Please login to your online account to display your discounted pricing

Amberlite™ IRP-69 ion-exchange resin, ACROS Organics™

$30.46 - $30.46

Chemical Identifiers

Chemical Name or Material Amberlite™ IRP-69
Assay 9.4 to 11.5% (Na)
MDL Number MFCD00132786
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC301311000

  SDS
thermo fisher scientific
301311000
Plastic bottle 100g Each for $30.46

AC301315000

  SDS
fisher scientific acros
301315000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100g
Assay 9.4 to 11.5% (Na)
Physical Form Powder
Particle Size Particle size analysis: screens on 100 Mesh: <1%, screens on 200 Mesh: 10.0 to 25.0%
Merck Index 14, 382
Ion Exchange Capacity potassium exchange capacity: 110 to 135mg/g
CAS 55464-99-8
Chemical Name or Material Amberlite™ IRP-69
MDL Number MFCD00132786
Ammonium Salts ammonium salts: Passes Test
Packaging Plastic bottle
Water 10% max. (K.F.)

Ion exchange resin