Please login to your online account to display your discounted pricing

Amberlite™ IRN-78 ion-exchange resin, OH-form, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC429295000

 View more versions of this product

Catalog No. AC429295000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Quantity 500g
Color Colorless to yellow
Chemical Name or Material Amberlite™ IRN-78 ion-exchange resin
Name Note Ion-exchange Resin, OH-form
MDL Number MFCD00145822
CAS 11128-95-3
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Health Hazard 1 Danger

Specifications

Chemical Name or Material Amberlite™ IRN-78 ion-exchange resin
Name Note Ion-exchange Resin, OH-form
CAS 11128-95-3
MDL Number MFCD00145822
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.

SDS & Safety Documents