Amberlite™ IRN-78, Amber beads, Alfa Aesar™
Strongly basic anion exchange resin
Manufacturer:  Alfa Aesar 45591A3
 View more versions of this product
Catalog No. AA45591A3
Specifications
Specifications
Amberlite™ IRN-78
Styrene-DVB
Amber
Beads
11128-95-3
MFCD00145822
2kg
Nuclear
Chemical Identifiers
11128-95-3
Styrene-DVB
MFCD00145822
SDS Documents