Amberlite™ IRN-77, ion exchange resin, Nuclear grade, Alfa Aesar™
Styrene-DVB; Strongly acidic cation exchange resin
Manufacturer:  Alfa Aesar 16821A3
 View more versions of this product
Catalog No. AA16821A3
Specifications
Specifications
Amberlite IRN-77
Brown beads
Styrene-DVB
1.1-1.4g/cm3
2kg
Nuclear
11128-94-2
Odorless
MFCD00145822
Brown
Beads
Chemical Identifiers
11128-94-2
Styrene-DVB
MFCD00145822
SDS Documents