Please login to your online account to display your discounted pricing

Amberlite™ IRN-150, ion exchange resin, Alfa Aesar™

Water treatment and purification

$129.89 - $129.89

Chemical Identifiers

Chemical Name or Material ion exchange resin
Molecular Formula Styrene-DVB
MDL Number MFCD00145822
Merck Index 14,382
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA40992A3

  SDS
alfa aesar
40992A3
Each for $129.89
Description & Specifications

Specifications

Quantity 2kg
Chemical Name or Material ion exchange resin
TSCA TSCA
Odor Odorless
Molecular Formula Styrene-DVB
MDL Number MFCD00145822
Merck Index 14,382