missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite™ IRC-76, ion exchange resin

Chemical Identifiers

CAS 11098-83-2
MDL Number MFCD00145802
Merck Index 14,382
Chemical Name or Material Amberlite™ IRC-76, Ion exchange resin, Acrylic-DVB; Weakly acidic gel type cation resin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3938436 SDS Alfa Aesar™
3938436
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39384A3 SDS Alfa Aesar™
39384A3
2kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

11098-83-2
MFCD00145802
14,382
Amberlite™ IRC-76, Ion exchange resin, Acrylic-DVB; Weakly acidic gel type cation resin

Specifications

11098-83-2
MFCD00145802
500g
Odorless
14,382
Amberlite™ IRC-76, Ion exchange resin, Acrylic-DVB; Weakly acidic gel type cation resin