Please login to your online account to display your discounted pricing

Amberlite™ IRC-748 ion-exchange resin, ACROS Organics™


Quantity 250g
Packaging Plastic Bottle

Manufacturer: fisher scientific acros  335392500

 View more versions of this product

Catalog No. AC335392500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 250g
Color Opaque
Merck Index 14, 382
Particle Size Particle size analysis: uniformity coefficient: <1.70,% <0.300mm: <1.0%,%>1.000mm: <5.0%, harmonic mean size: 0.500 - 0.650mm
Chemical Name or Material Amberlite™ IRC-748
Moisture Holding Capacity (Na+): 60 to 65%
Additional Information volume cation capacity (Na+):>1.35 eq/L
Packaging Plastic Bottle
MDL Number MFCD00132702
CAS 79620-28-3

Specifications

Chemical Name or Material Amberlite™ IRC-748
CAS 79620-28-3
MDL Number MFCD00132702