missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amberlite™ IRA-900 Cl ion-exchange resin, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202315000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resinSpecifications
Beads | |
9050-97-9 | |
Plastic bottle | |
C4H12ClN | |
(Cl-): 58 to 64% | |
01,511; 02,267; 04,267; 06,302; 07,182 | |
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl | |
[Cl-].C[N+](C)(C)C | |
109.60 | |
CHEBI:7070 |
Amberlite™ IRA-900 | |
Particle size analysis : uniformity coefficient: <1.8, harmonic mean size: 0.650 to 0.820mm, <0.300mm: <0.5%,> 1.180mm: <3.0% | |
(Cl-):> 1.0 eq/L | |
MFCD00132712 | |
Beige/Brown to Ivory | |
500 g | |
OKIZCWYLBDKLSU-UHFFFAOYSA-M | |
tetramethylazanium;chloride | |
6379 |
Chemical Identifiers
9050-97-9 | |
109.60 | |
OKIZCWYLBDKLSU-UHFFFAOYSA-M | |
6379 | |
[Cl-].C[N+](C)(C)C |
C4H12ClN | |
MFCD00132712 | |
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl | |
CHEBI:7070 |
RUO – Research Use Only