Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite IRA-410(Cl), ion exchange resin

$34.18 - $361.73

Chemical Identifiers

Chemical Name or Material Amberlite IRA-410(Cl)
CAS 9002-26-0
Name Note ion exchange resin
MDL Number MFCD00132710
Merck Index 14,382
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA177340E

 
alfa aesar™
A177340E
2500g Each for $361.73

AAA1773422

 
alfa aesar™
A1773422
100g Each for $34.18

AAA1773436

 
alfa aesar™
A1773436
500g Each for $95.43
Description & Specifications

Specifications

TSCA TSCA
Odor Amine-like
Name Note ion exchange resin
MDL Number MFCD00132710
Merck Index 14,382
Quantity 2500g
Chemical Name or Material Amberlite IRA-410(Cl)
CAS 9002-26-0