Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite IRA-402(OH), ion exchange resin

$72.80 - $310.93

Chemical Identifiers

Chemical Name or Material Amberlite IRA-402(OH)
CAS 52439-77-7
Name Note ion exchange resin
MDL Number MFCD00145564
Merck Index 14,382
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1818522

 
alfa aesar™
A1818522
100g Each for $72.80

AAA1818536

 
alfa aesar™
A1818536
500g Each for $310.93
Description & Specifications

Specifications

Chemical Name or Material Amberlite IRA-402(OH)
Name Note ion exchange resin
MDL Number MFCD00145564
CAS 52439-77-7
Merck Index 14,382
Quantity 100g

Applications
Amberlite IRA-402(OH) ion exchange resin is a strongly basic gel-type anion exchange resin. It is used to adsorb uranium(VI) from sulphate, removal of cyanide and zinc cyanide. It is also used to produce perchlorate and the free nucleoside. Further, it is used for water conditioning.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and acids.