Please login to your online account to display your discounted pricing

Amberlite™ IRA-400(Cl), ion exchange resin, Alfa Aesar™

$30.14 - $271.22

Chemical Identifiers

Chemical Name or Material Amberlite IRA-400, ion exchange resin
CAS 9002-24-8
Molecular Formula Styrene-DVB; Strongly basic gel type resin
MDL Number MFCD00132708
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1724636

  SDS
alfa aesar
1724636
500g Each for $87.39

AA1724622

  SDS
alfa aesar
1724622
100g Each for $30.14

AA17246A3

  SDS
alfa aesar
17246A3
2kg Each for $271.22
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Amberlite IRA-400, ion exchange resin
Molecular Formula Styrene-DVB; Strongly basic gel type resin
MDL Number MFCD00132708
CAS 9002-24-8