Amberlite™ IRA-400(Cl), ion exchange resin, Alfa Aesar™
Manufacturer:  Alfa Aesar 17246A3
 View more versions of this product
Catalog No. AA17246A3
Specifications
Specifications
Amberlite IRA-400, ion exchange resin
Styrene-DVB; Strongly basic gel type resin
2kg
9002-24-8
MFCD00132708
Chemical Identifiers
9002-24-8
Styrene-DVB; Strongly basic gel type resin
MFCD00132708
SDS Documents