Please login to your online account to display your discounted pricing

Amberlite™ IR120, H form, ion-exchange resin, ACROS Organics™

$55.41 - $176.62

Chemical Identifiers

Chemical Name or Material Amberlite™ IR120
CAS 78922-04-0
MDL Number MFCD00132707
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369720051

  SDS
thermo fisher scientific
AC369720051
Plastic Bucket 5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC369720010

  SDS
fisher scientific acros
369720010
Plastic bottle 1kg Each for $176.62

AC369722500

  SDS
fisher scientific acros
AC369722500
Plastic bottle 250g Each for $55.41

AC369720050

  SDS
thermo fisher scientific
AC369720050
Plastic bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Quantity 5kg
Color Amber to yellow-brown
Packaging Plastic Bucket
Particle Size Particle size analysis: uniformity coefficient <1.8, harmonic mean size: 0.620 to 0.830mm,% <0.300mm: <2.0%,%>1.180mm: <3.0%
Moisture Holding Capacity (H+): 53.0 to 58.0%
Volume Total Capacity (H+):> 1.8 eq/L
Chemical Name or Material Amberlite™ IR120
CAS 78922-04-0
MDL Number MFCD00132707

Ion exchange resin