Please login to your online account to display your discounted pricing

Amberlite™ IR120, H form, ion-exchange resin, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC369722500

 View more versions of this product

Catalog No. AC369722500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 250g
Color Amber to yellow-brown
Particle Size Particle size analysis: uniformity coefficient <1.8, harmonic mean size: 0.620 to 0.830mm,% <0.300mm: <2.0%,%>1.180mm: <3.0%
Moisture Holding Capacity (H+): 53.0 to 58.0%
Packaging Plastic bottle
Volume Total Capacity (H+):> 1.8 eq/L
CAS 78922-04-0
Chemical Name or Material Amberlite™ IR120
MDL Number MFCD00132707

Specifications

Chemical Name or Material Amberlite™ IR120
CAS 78922-04-0
MDL Number MFCD00132707