Please login to your online account to display your discounted pricing

Amberlite™ IR-120 Na ion-exchange resin, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  208412500

 View more versions of this product

Catalog No. AC208412500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 250g
Color Amber to brown
Merck Index 14, 382
Particle Size Particle size analysis: uniformity coefficient: <1.9, harmonic mean size: 0.600 to 0.800mm,% <0.300mm: <2.0%
Fieser 01,511; 04,266; 05,355; 06,302; 09,256
Volume Total Capacity (Na+):> 2.0 eq/L
Packaging Plastic bottle
MDL Number MFCD00132707
Moisture Holding Capacity (Na+): 45 to 50%
Chemical Name or Material Amberlite™ IR120
CAS 9002-23-7

Specifications

Chemical Name or Material Amberlite™ IR120
CAS 9002-23-7
MDL Number MFCD00132707