Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite™ IR-120(H), ion exchange resin


Quantity 250g

Manufacturer: alfa aesar™  L1428530

 View more versions of this product

Catalog No. AAL1428530

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Amberlite™ IR-120(H)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Name Note ion exchange resin
Merck Index 14,382
MDL Number MFCD00132707
Health Hazard 3 GHS P Statement
P280i-P305+P351+P338-P310
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
CAS 78922-04-0
Quantity 250g

Specifications

Chemical Name or Material Amberlite™ IR-120(H)
CAS 78922-04-0
Name Note ion exchange resin
MDL Number MFCD00132707
Merck Index 14,382

Applications
Amberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Accessories

Danger

GHS P Statement
P280i-P305+P351+P338-P310
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.

SDS & Safety Documents