Please login to your online account to display your discounted pricing

Amberlite™ FPC3500, ion-exchange resin, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC389992500

 View more versions of this product

Catalog No. AC389992500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 250g
Color Light yellow
Density 1.3500g/mL
Moisture Moisture hold cont.(H):60.0 to 70.0%
Particle Size Uniformity coefficient:max. 2.00, Harmonic mean size:0.450 to 0.650mm, Particle size (<0.300mm): max. 2.0%, (>1.180mm): max. 5.0%
Packaging Plastic bottle
MDL Number MFCD08460528
CAS 950182-46-4
CAS 7732-18-5
Chemical Name or Material Amberlite™ FPC3500
Additional Information Total exchange capacity (H):min. 2.60 eq/L

Specifications

Chemical Name or Material Amberlite™ FPC3500
CAS 950182-46-4
CAS 7732-18-5
MDL Number MFCD08460528