Please login to your online account to display your discounted pricing

Alfa Aesar™ Amberlite™ FPC3500(H), ion exchange resin

$59.51 - $59.51

Chemical Identifiers

Chemical Name or Material Amberlite™ FPC3500 (H) ion exchange resin
MDL Number MFCD08460528
Merck Index 14,832
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4590330

  SDS
alfa aesar™
4590330
250g Each for $59.51
Description & Specifications

Specifications

Chemical Name or Material Amberlite™ FPC3500 (H) ion exchange resin
Density 1.2g/mL
MDL Number MFCD08460528
Color White
Merck Index 14,832
Quantity 250g

  • Suitable for the neutralization of strong bases, the recovery of metallic ions, and the isolation and concentration of antibiotics, basic amino acids, enzymes, and peptides