missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals

$118.94 - $1,265.37

Chemical Identifiers

CAS 9042-11-9
MDL Number MFCD00132693
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC202110250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
202110250
25 g Plastic bottle
Each for $118.94
 
AC202115000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
202115000
500 g Plastic bottle
Each for $1,265.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
Specifications

Chemical Identifiers

9042-11-9
MFCD00132693

Specifications

9042-11-9
MFCD00132693
14, 382
Amberlite™ CG-50 - type 1
Plastic bottle
01,511; 04,266; 05,355; 06,302
25 g
Safety and Handling

Safety and Handling

Recommended Storage : Indoors +20°C

SDS

RUO – Research Use Only