missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amberjet™ 4200-Cl, Acros Organics

$119.90 - $119.90

Chemical Identifiers

CAS 60177-39-1
MDL Number MFCD00145586
Chemical Name or Material Amberjet™ 4200-Cl
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC301330025 SDS Thermo Fisher Scientific
AC301330025
2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC301335000 SDS Thermo Fisher Scientific
301335000
Glass Bottle 500g Each for $119.90
Specifications Description & Specifications

Chemical Identifiers

60177-39-1
MFCD00145586
Amberjet™ 4200-Cl

Specifications

Beads
Yellow translucent
MFCD00145586
(Cl): 49% to 55%
Amberjet™ 4200-Cl
2.5kg
0.600 to 0.800mm harmonic mean size, 0.5% Max. (<0.425mm), 5.0% Max. (>0.850mm), 1.25 uniformity coefficient Max.
(Cl): 1.30 eq/L min.
60177-39-1