Please login to your online account to display your discounted pricing

Amberjet™ 4200-Cl, Acros Organics


Quantity 500g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  301335000

 View more versions of this product

Catalog No. AC301335000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 500g
Color Yellow translucent
Particle Size 0.600 to 0.800mm harmonic mean size, 0.5% Max. (<0.425mm), 5.0% Max. (>0.850mm), 1.25 uniformity coefficient Max.
Chemical Name or Material Amberjet™ 4200-Cl
Moisture Holding Capacity (Cl): 49% to 55%
Packaging Glass Bottle
Volume Total Capacity (Cl): 1.30 eq/L min.
MDL Number MFCD00145586
CAS 60177-39-1

Specifications

Chemical Name or Material Amberjet™ 4200-Cl
CAS 60177-39-1
MDL Number MFCD00145586