Fisherbrand™ Amber Wide Mouth Packer Bottles with White Polypropylene Pulp/Vinyl Cap
Ideal general purpose bottle for solids. Fisherbrand™ Amber Wide Mouth Packer Bottles with White Polypropylene Pulp/Vinyl Cap are often used in chemical and environmental applications.
$35.60 - $930.57
Specifications
Liner Material Pulp/Vinyl
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Amber
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-911-435A Fisherbrand™
ALP263518
45-400 24/Cs. 8.5 oz. 250mL Case of 24 Each for $57.23
02-911-450A Fisherbrand™
ALP263519
45-400 84/Cs. 8.5 oz. 250mL Case of 84 Each for $134.67
02-911-432A Fisherbrand™
ALP263520
28-400 24/Cs. 1 oz. 30mL Case of 24 Each for $36.00
02-911-436A Fisherbrand™
ALP263521
53-400 12/Cs. 16 oz. 500mL Case of 12 Each for $48.67
02-911-434A Fisherbrand™
ALP263517
38-400 24/Cs. 4 oz. 120mL Case of 24 Each for $35.60
02-911-433A Fisherbrand™
ALP263524
33-400 24/Cs. 2 oz. 60mL Case of 24 Each for $38.40
02-911-437A Fisherbrand™
ALP263527
53-400 12/Cs. 32 oz. 950mL Case of 12 Each for $71.40
02-911-448A Fisherbrand™
ALP263523
33-400 216/Cs. 2 oz. 60mL Case of 216 Each for $271.20
02-911-449A Fisherbrand™
ALP263516
38-400 180/Cs. 4 oz. 120mL Case of 180 Each for $207.90
02-911-452A Fisherbrand™
ALP263528
53-400 36/Cs. 32 oz. 950mL Case of 36 Each for $155.53
02-911-799A Fisherbrand™
ALP263515
28-400 624/Cs. 0.50 oz. 15mL Case of 624 Each for $656.10
02-911-852A Fisherbrand™
ALP263525
20-400 720/Cs. 0.2 oz. 6mL Case of 720 Each for $930.57
02-911-857A Fisherbrand™
ALP263526
20-400 912/Cs. 0.27 oz. 8mL Case of 912 Each for $720.03
02-911-451A Fisherbrand™
ALP263522
53-400 60/Cs. 16 oz. 500mL Case of 60 Each for $215.87
Specifications
Description
  • Features wide mouth opening for easy transfer of contents
  • Amber color filters out ultraviolet wavelengths, ideal for light sensitive solids
  • Classic “vitamin bottles” and are often used in pharmaceutical packaging
Poplypropylene Pulp/Vinyl Lined Caps
  • Good impact strength and pliability 
  • Can be remelted after they are formed
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Description & Specifications
Specifications
Pulp/Vinyl
Screw
Amber
Wide
Polypropylene
White
Glass
Round