missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Custom Cleaned Amber Boston Round Bottles

Ideal storage for solvents, chemicals and samples. Fisherbrand™ Custom Cleaned Amber Boston Round Bottles are manufactured from amber colored soda glass.

Manufacturer:  Fisherbrand™ 02912002

 View more versions of this product

Catalog No. 02-912-002



Description

Description

  • Narrow mouth
  • Pre-cleaned for visible particulates using deionised water
  • With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications

Specifications

8.45 oz., 250mL
8.45 oz.
Amber
12/PK.
White
Polyethylene
24-400 mm
Screw Cap
Round
250 mL
USP purified Water
Glass
Boston Round Bottle
PTFE
Yes
PTFE
Wide
SDS
Documents
Product Certifications