Please login to your online account to display your discounted pricing
Qorpak™ Amber Glass Jugs, Ready to Clean
Designed to make this readily available for cleaning process
$124.66 - $124.66
Specifications
Closure Liner PTFE
Closure Material Polypropylene
Closure Screw
Closure Color Black
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Quantity Mouth Price Quantity    
02-992-061
qorpak™
GLC-02058
84 oz., 2500mL 12.2 in. 6/Cs. Wide Case for $124.66
Description & Specifications
Specifications
Closure Liner PTFE
Closure Material Polypropylene
Closure Screw
Closure Color Black
Color Amber
Material Glass
Shape Cylindrical
  • Ideal for light-sensitive products to protect against UV radiations
  • Molded finger grips facilitates easy handling and makes this perfect for storage of liquids in large quantity
  • With polypropylene caps and PTFE disc attached