Please login to your online account to display your discounted pricing

Kimble™ Amber Glass Dropping Bottles


For storage of light-sensitive chemicals.

Manufacturer: kimble™  K15040G30*

 View more versions of this product

Catalog No. S31683

  • / Pack of 12 Each
  • / Case of 6 Pack
 

Description & Specifications

Specifications

Capacity (Metric) 30mL
Capacity (English) 1 oz.
Closure Screw
Closure Material Polypropylene
Color Amber
Material Glass
Capacity 1 oz., 30mL