Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps
For light sensitive solvents, chemicals, and samples. Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps are general use bottles for long term storage of UV sensitive contents.
$31.07 - $263.00
Specifications
Liner Material Pulp/Vinyl
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Amber
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-911-723A Fisherbrand™
ALP263435
22-400 160/Cs. 4 oz. 120mL Case of 160 Each for $200.57
02-911-725A Fisherbrand™
ALP263424
28-400 60/Cs. 16 oz. 480mL Case of 60 Each for $147.10
02-911-724A Fisherbrand™
ALP263438
24-400 108/Cs. 8 oz. 240mL Case of 108 Each for $187.07
02-911-729A Fisherbrand™
ALP263433
33-400 30/Cs. 32 oz. 960mL Case of 30 Each for $130.37
02-911-742A Fisherbrand™
ALP263429
20-400 288/Cs. 2 oz. 60mL Case of 288 Each for $263.00
02-911-747A Fisherbrand™
ALP263432
33-400 12/Cs. 32 oz. 960mL Case of 12 Each for $55.83
02-911-745A Fisherbrand™
ALP263423
28-400 12/Cs. 16 oz. 480mL Case of 12 Each for $36.60
02-911-746A Fisherbrand™
ALP263428
20-400 24/Cs. 2 oz. 60mL Case of 24 Each for $31.07
02-911-748A Fisherbrand™
ALP263436
22-400 24/Cs. 4 oz. 120mL Case of 24 Each for $41.20
02-911-749A Fisherbrand™
ALP263439
24-400 12/Cs. 8 oz. 240mL Case of 12 Each for $34.13
02-912-337A Fisherbrand™
ALP263426
20-400 48/Cs. 1 oz. 30mL Case of 48 Each for $49.27
Specifications
Description
Amber Boston Round Bottles
  • Boston round bottles features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber glass bottles absorb all wavelengths in the ultraviolet spectrum
White Polypropylene Pulp/Vinyl Lined Caps
  • Good impact strength, thermal resistance and pliability
  • Autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Description & Specifications
Specifications
Pulp/Vinyl
Screw
Amber
Narrow
Polypropylene
White
Glass
Round