Please login to your online account to display your discounted pricing
Qorpak™ Amber Boston Round Bottles with Black Phenolic Pulp/Tin Foil Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic Pulp/Tin Foil Cap ensures product integrity for long-term storage.
$93.06 - $93.06
Specifications
Closure Liner Pulp/Tin Foil
Closure Material Phenolic
Closure Color Black
Closure Type Screw
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Type Quantity Closure Size Price Quantity    
02-992-568
qorpak™
GLC01915
2 oz., 60mL Convention pack 24/Cs. 20-400 Case of 24 Each for $93.06
Description & Specifications
Specifications
Closure Liner Pulp/Tin Foil
Closure Material Phenolic
Closure Color Black
Closure Type Screw
Color Amber
Material Glass
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum