Please login to your online account to display your discounted pricing

Ambenonium dichloride, Tocris

Cholinesterase inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Ambenonium dichloride
Assay Percent Range >99%
Molecular Formula C28H42Cl4N4O2
Formula Weight 608.48
Synonym N,N'-[(1,2-Dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)]bis(2-chloro-N,N-diethylbenzenemethaminium) dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

038850

 
r & d systems
RDS038850
50mg Each for $682.67

038810

 
r & d systems
RDS038810
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 608.48
Chemical Name or Material Ambenonium dichloride
CAS 52022-31-8
Molecular Formula C28H42Cl4N4O2
Assay Percent Range >99%
Synonym N,N'-[(1,2-Dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)]bis(2-chloro-N,N-diethylbenzenemethaminium) dichloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature