Please login to your online account to display your discounted pricing

AM 580, Tocris

Retinoic acid analog; RARα agonist

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material AM 580
Assay Percent Range >98%
Molecular Formula C22H25NO3
Formula Weight 351.44
Synonym 4-[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carboxamido]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

076010

 
r & d systems
RDS076010
10mg Each for $195.49

076050

 
r & d systems
RDS076050
50mg Each for $816.90
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 351.44
Assay Percent Range >98%
Synonym 4-[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carboxamido]benzoic acid
Molecular Formula C22H25NO3
CAS 102121-60-8
Chemical Name or Material AM 580
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature