Please login to your online account to display your discounted pricing

AM 580, Tocris


Retinoic acid analog; RARα agonist

Quantity 10mg

Manufacturer: r & d systems  RDS076010

 View more versions of this product

Catalog No. 076010

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 351.44
Chemical Name or Material AM 580
Assay Percent Range >98%
Molecular Formula C22H25NO3
CAS 102121-60-8
Synonym 4-[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carboxamido]benzoic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material AM 580
Assay Percent Range >98%
Molecular Formula C22H25NO3
Formula Weight 351.44
Synonym 4-[(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)carboxamido]benzoic acid
Accessories

Store at Room Temperature