Please login to your online account to display your discounted pricing

AM 404, Tocris


Anandamide transport inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS111610

 View more versions of this product

Catalog No. 111610

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 395.58
Quantity 10mg
Chemical Name or Material AM 404
Molecular Formula C26H37NO2
CAS 198022-70-7
Assay Percent Range >98%
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Recommended Storage Store at -20°C

Specifications

Chemical Name or Material AM 404
Assay Percent Range >98%
Molecular Formula C26H37NO2
Formula Weight 395.58
Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Accessories

Store at -20°C